Customize ToolTips - Customize ToolTips - Proteome Discoverer 3.1 User Guide - Proteome Discoverer Software - Customize ToolTips - Proteome Discoverer Software - Proteome Discoverer Software - topic
Proteome Discoverer 3.1 User Guide
- Product Tree
- Ardia Platform > Software > Proteome Discoverer Software
- Desktop Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- Right-click the empty space in the toolbar and select Customize.
- The Customize dialog box opens.
- Select the Options tab.
- To display ToolTips when you point to the icons, select the Show ScreenTips on Toolbars option.
- (Optional) To display shortcut keys with the ToolTips, select the Show Shortcut Keys in ScreenTips option.
- You must select Show ScreenTips on Toolbars to use the Show Shortcut Keys in ScreenTips option.
- To save your changes and close the Customize dialog box, select Close.