Modify icons and menu commands - Modify icons and menu commands - Proteome Discoverer 3.1 User Guide - Proteome Discoverer Software - Modify icons and menu commands - Proteome Discoverer Software - Proteome Discoverer Software - topic
Proteome Discoverer 3.1 User Guide
- Product Tree
- Ardia Platform > Software > Proteome Discoverer Software
- Desktop Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- Right-click the empty space in the toolbar and select Customize.
- The Customize dialog box opens.
- Select the Commands tab.
- Select the icon or menu command.
- Select Modify Selection on the Commands page.
- Select an option.
- Refer to the Help in any Microsoft Office product for information about available options.
- Select Close.