The following table describes the parameters on the Fragment Match Options page.
Command or Option | Description |
|---|---|
Loads the last saved default fragment match settings for the selected mass analyzer/activation type combination. | |
Saves the current fragment match settings as the default fragment match settings for the selected mass analyzer/activation type combination. | |
Loads the original factory fragment match settings for all mass analyzer/activation type combinations. | |
Charge Detail Level | Selects the target charge state to display from the following settings:
|
Annotation Threshold | Specifies a peak intensity limit. Peaks below this limit are not annotated. This limit prevents the addition of too many labels to a spectrum, especially if the low-intensity peaks are possibly noise peaks.
Default: 1.0
Default: 1000 |
Show Legend | Determines whether a legend consisting of colored lines that denote the displayed ion series appears at the bottom of each spectrum image. |
Show Reference Spectrum | Determines whether to display the reference spectrum from the reference spectrum library. |
Ref. Spectrum Source |
|
Use Search Settings | Annotates the spectra using the match tolerances and ion series settings that you chose for the search node. |
Mass Analyzer | Specifies which mass analyzer was used to obtain the spectrum. |
Match Tolerance | Specifies the mass tolerance. The application annotates the spectrum with ions that it assigns within this mass tolerance. You can use daltons or millimass units. |
Activation Type | Specifies what type of activation produced the fragments. |
Ion Series | Selects which types of ion series to annotate in the spectra. |
Neutral Losses | Selects which types of neutral losses to annotate in the spectra. |
Other | Annotates the spectra with immonium ions or precursor ions. |
Abbreviations |
|