You can access context-sensitive Help for information about a command.
Procedure
  1. Right-click the empty space in the toolbar and choose Customize.
  2. The Customize dialog box opens.
  3. Select the Commands tab.
  4. In the Commands list, select the command that you want information about.
  5. Select Description.