The following table describes the job queue icons and column headings.
Parameter | Description |
|---|---|
Stops a search. | |
Removes a search. | |
Selects that job to be run immediately after the current run finishes. | |
Refreshes the display of the job queue. | |
Opens a completed results report (result file) from the selected search. | |
Opens a study (.pdStudy file) from the selected search. | |
Displays any longer messages issued during processing. | |
Execution State | Displays the status of the search:
|
Details | Indicates whether there were warnings or errors in the workflow. |
Progress | Displays job completion as a percentage. |
Type | Displays the type of workflow: Processing or Consensus. |
Name | Displays the name of your project from the name that you specified in the Workflow Editor. |
Submitted at | Displays the date and time that you submitted the search. |
Study | Displays the name of the study that the search belongs to. |
Data Source | Displays the path and name of the raw data file being used in the search. |
Description | Displays the additional descriptive information that you entered when you used the Workflow Editor. Otherwise, this field is blank. |
Time | Displays the time that the processing finished for each step described in the Message column. |
Processing Node | Displays the name of the workflow node performing the step described in the Message column. |
Level | Displays the type of information that the adjacent message contains. By default, some of the types are not visible, so you must use the Custom column filter to select them. This column can display these message types:
|
Message | Displays the nature of each step in the processing. |