The job queue displays four types of messages in the Type column as it processes a workflow:
- Info: For short informational messages
- Verbose: For long informational messages
- Warning: For warning messages
- Error: For error messages
By default, the job queue displays all messages except Verbose messages. However, you can display Verbose messages.
Procedure
- In the job queue toolbar, select the Display Verbose Messages check box,
.
- If you do not immediately see it, click the double arrow,
, in the upper right of the job queue window.
- The job queue displays any Verbose messages and distinguishes them from shorter informational messages by displaying “Verbose” in the Type column.
Verbose messages in the job queue - If a verbose message is longer than the width of the line, point to the message in the Message column.
- The full text of the message appears in a tooltip.