The Spectrum Exporter node converts the spectrum from the available input formats, such as RAW and MGF files, to MGF, MZDATA, or MZML output files for processing elsewhere.
When you use the Spectrum Exporter with the Spectrum Selector node, the Spectrum Grouper node, or both, you can use the results with additional or customized data processing that is required outside of the application.
The following table describes the parameters for the Spectrum Exporter node.
Parameter | Definition |
|---|---|
File Name | Specifies the name of the output file that contains the exported data. |
File Name Suffix | Appends a suffix to the output file name. When you use multiple export nodes in a workflow—for example, to export HCD and ETD spectra—you can use the suffix to distinguish the contents of each output file. |
Export Format | Specifies the format of the exported spectra:
For instructions, see Export Spectra. |