Specify a value for the Maximum Rank parameter to set a threshold that determines which PSMs the application transfers to the report file.
You can set the threshold to any positive value. The default is 0, which means that the Maximum Rank filter is not applied.
The application transfers to the report file only those PSMs with a rank better than or equal to the specified threshold.