Procedure
  1. Click Show Advanced Parameters in the Parameters pane of the Workflow Editor, and specify a value for the Maximum Delta Cn parameter to set a threshold that determines which PSMs the application transfers to the report file.
The threshold can be within a range of 0 to 0.1. The default is 0.05.
The application transfers only those PSMs with a Δ Cn value greater than the specified threshold to the report file.
MSF Files node parameters