The following table describes the parameters on the Find Protein References page of the FASTA Database Utilities dialog box.
Parameters | Description |
|---|---|
FASTA Database | Specifies the name of the FASTA file that you want to search for sequences or references in. |
Search For | Specifies the amino acid sequence to search for. |
Search In | Determines whether to search for the search string in the protein references or sequences.
|
Maximum Number of Matches Reported | Specifies the maximum number of matching references or sequences to report. |
Start Search | Begins the search for the reference or sequence. |
Save/Add Selected to Database | Adds a selected protein result row to a specified database. |
Stop Search | Suspends the active search. |
Reference: List of Operators | Specifies the operator for the condition:
|
Reference: List of Condition Settings | Custom: Opens the Enter Filter Criteria for Reference dialog box so that you can define additional conditions. |
Clear Reference Filter icon ( | Deletes the condition under “Reference” on the Find Protein References page. |
Clear All Filters | Deletes all conditions in the Enter Filter Criteria for Reference dialog box and the condition under “Reference” on the Find Protein References page. |