The following table describes the parameters on the Compile FASTA Database page of the FASTA Database Utilities dialog box.
Parameters | Description |
|---|---|
Original | Specifies the path and name of the source FASTA database that you are taking the information from. It must be an existing FASTA file. |
Target | Specifies the path and name of the FASTA file that you are placing or adding the extracted information to. |
Target Database Options | Determines whether the extracted information is placed in a new FASTA file or is appended to an existing FASTA file.
|
Search In | Determines whether to search for the search string in the protein references or sequences.
|
Ignore Case of Reference Strings | Disregards the case of the text in the Step 1: String(s) to Include box and the Step 2: String(s) to Exclude From the Results of Step 1 box. |
Step 1: String(s) to Include: | Specifies the information to be extracted from the original file. |
Active | Specifies the number of the condition set and allows you to select it. |
Select Operator | Specifies the action to apply to the condition selected in the Condition column:
|
Condition | Specifies the condition that the information must meet in order to be extracted. |
Adds a line to the Step 1: String(s) to Include box. | |
Deletes a line from the Step 1: String(s) to Include box. | |
Step 2: String(s) to Exclude From the Results of Step 1 | Specifies the information to be excluded from the extracted results. |
Active | Specifies the number of the condition set and allows you to select it. |
Select Operator | Specifies the action to apply to the condition selected in the Condition column:
|
Condition | Specifies the condition that the information must meet in order to be excluded. |
| Adds a line to the Step 2: String(s) to Exclude From the Results of Step 1 box. |
| Deletes a line from the Step 2: String(s) to Exclude From the Results of Step 1 box. |
Compile Database | Compiles and saves the changes to the FASTA file specified in the Target box. |
Stop | Suspends the compilation. |