The following table describes the parameters on the Add Protein References page of the FASTA Database Utilities dialog box.
Parameters | Description |
|---|---|
FASTA File | Specifies the name of the FASTA file to modify. |
Enter Description | Describes the protein sequence that you are appending to the selected FASTA file. Include the name of the protein and the source tissue. |
Enter Protein Sequence | Specifies the amino acid sequence of the protein that you are appending to the selected FASTA file. |
Add Entry | Appends and saves the protein sequence and reference to the currently selected FASTA file. |