If you obtain unexpected quantification results, verify that all node settings in your processing workflow are reasonable.
- Make sure that the Integration Tolerance parameter of the Reporter Ions Quantifier node fits the data that you are processing. The default is 20 ppm, which is too low if you are processing PQD data from an ion trap.
- Make sure that the settings of the Mass Analyzer, MS Order, and Activation Type parameters of the Reporter Ions Quantifier node are correct for the data that you are processing. The following figure shows the typical settings to use for quantifying HCD scans from the Orbitrap mass spectrometer.
To quantify PQD scans from an ion trap, use the typical settings.
Make sure that you use the correct set of static and dynamic modifications for the search engine. For example, if you are searching iodo TMT 6plex data with Sequest HT, check that your settings resemble those in the following figure.