Procedure
- If the processing method that you want to modify is not open, open it and make sure that the parameter file and location are specified as described in Create a processing method that calls the Daemon utility from the Xcalibur data system.
- In the Std, QC, Unk, and Other columns, do the following:
- To send a sample to the Daemon utility, make sure that “Yes” appears in the column for its sample type.
- To avoid processing a sample with the utility, clear the column for its sample type.
TIP
Use the Other column for the Blank sample type. For example, if you do not want to send blank samples to the utility for further processing, clear the Other column.
- Save the processing method.