Display the Enrichment Chart - Display the Enrichment Chart - Proteome Discoverer 3.1 User Guide - Proteome Discoverer Software - Display the Enrichment Chart - Proteome Discoverer Software - Proteome Discoverer Software - topic
Proteome Discoverer 3.1 User Guide
- Product Tree
- Ardia Platform > Software > Proteome Discoverer Software
- Desktop Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- Open the result of interest.
- Select the Proteins tab.
- Select the Enrichment Chart,
, in the toolbar, or choose View > Enrichment Chart. - Select one or more entries in the Proteins list.