The shortcut menu in the File Alignment view provides options to copy or export the plot as an image or the underlying data.
Command or Option | Description |
|---|---|
Show Position Tooltips | Displays the retention time of the selected file and the delta retention time of the same landmark in the reference file. |
Zoom Out | Reduces the view. |
Undo All Zoom/Pan | Returns the Retention Tine Alignment view to the default view. |
Copy | Copies the data in the File Alignment view. |
Image | Copies the data in the File Alignment view as an image. |
Points | Copies the data in the File Alignment view as text. |
Export | Exports the data in the File Alignment view to a file. |
Image As | Exports the data in the File Alignment view as an image in an Enhanced Metafile (EMF) file. |
Points As | Exports the data in the File Alignment view as text in a TXT file. |