The following table describes the commands in the shortcut menu of the PCA Plots view.
Command or Option | Description |
|---|---|
Show Position Tooltips | Displays the following information:
|
Zoom Out | Reduces the view. |
Undo All Zoom/Pan | Returns the PCA plot to the default view. |
Copy | Copies the data in the plot view. |
Image | Copies the data in the plot view as an image. |
Points | Copies the data in the plot view as text. |
Export | Exports the data in the plot view to a file. |
Image As | Exports the data in the plot view as an image in an Enhanced Metafile (EMF) file. |
Points As | Exports the data in the plot view as text in a TXT file. |
Show Legend | Displays the meaning of the colors of groups of proteins or peptide groups. |