The following table describes the commands in the shortcut menu of the Volcano Plots view.
Command or Option | Description |
|---|---|
Show Position Tooltips | Displays the following information:
|
Zoom Out | Reduces the view. |
Undo All Zoom/Pan | Returns the volcano plot to the default view. |
Copy | Copies the data in the plot view. |
Image | Copies the data in the plot view as an image. |
Points | Copies the data in the plot view as text. |
Point Details | Copies all items on the selected page (Proteins or Peptide Groups) into a CSV-format text file. |
Export | Exports the data in the plot view to a file. |
Image As | Exports the data in the plot view as an image in an Enhanced Metafile (EMF) file. |
Points As | Exports the data in the plot view as text in a TXT file. |
Point Details As | Exports all items on the selected page (Proteins or Peptide Groups) into a CSV-format text file. |
Show Legend | Displays the meaning of the colors of groups of proteins or peptide groups. |
Check Point | Selects a data point in the plot view, colors the point blue, and selects the checkbox of the corresponding protein or peptide group in the Checked column of the result report. |
Uncheck Point | Clears a data point in the plot view and returns the point to its original color in the plot. In the Checked column of the result report, the application clears the checkbox of the corresponding protein or peptide group. |
Check All Visible Points | Selects all data points in the plot view, colors the points blue, and selects the checkboxes of the corresponding proteins or peptide groups in the Checked column of the result report. |
Uncheck All Visible Points | Clears all data points in the plot view, returns the points to their original colors, and re-displays the legend. In the Checked column of the result report, the application clears the checkboxes of the corresponding proteins or peptide groups. |