The following table describes the columns on the Quan Spectra page.
Parameters | Description |
|---|---|
# PSMs | Displays the total number of PSMs attached to the quantification spectrum. |
Abundances | Displays the abundances of the quantification channels. |
Activation Type | Displays the fragmentation method used to produce the product spectrum. |
Average Reporter S/N | Displays the average reporter S/N values, which the application calculates as the sum of S/N values found, divided by the number of defined tags. |
Best PSM Ambiguity | Displays the “best” ambiguities of the PSMs attached to a spectrum. This column can display the following categories:
For more information on the protein group inference process that the application uses to assign the grouping status of PSMs, see Understanding protein grouping. |
Checked | Indicates whether the item is selected. |
Creator Node No | Displays the number of the node in the workflow that created the spectrum. |
File ID | Displays a short identifier of the input file that the spectrum was taken from. |
First Scan | Displays the number of the first scan in grouped scans used to produce the composite search input scan, if grouping is performed. |
Ion Inject Time [ms] | Specifies the time in milliseconds used to accumulate ions in the mass spectrometer to reach their target value before they are scanned out. Low ion injection times are desirable because they indicate high transmission and fast cycle times. Ion injection times that slowly increase might indicate lower transmission, the need for multiplier gain calibration, or both. The MS/MS Spectrum page displays the Ion Inject Time [ms] column by default for every searched spectrum and every PSM for newly processed data. |
Isolation Interference [%] | Displays the percentage of interference by coisolation within the precursor isolation window. The application calculates the isolation interference only for precursors from MS1 spectra with high resolution. For more information on coisolation, see Excluding PSMs with high levels of coisolation. |
Last Scan | Displays the number of the last scan in grouped scans used to produce the composite search input scan, if grouping is performed. The last scan might not be the same as the first scan if the workflow enabled grouped scans. |
Mass Analyzer | Displays the mass analyzer used to create the raw data file. The application recognizes the following mass analyzers:
|
Master Scan(s) | Displays the master scans of the scans that the application used to derive the actual scan, usually one MS1 scan. |
MS Order | Indicates the order of the MS spectrum:
|
Original Mass | Displays the original mass of the precursor. |
Original Precursor Charge | Displays the charge state of the precursor as stored in the scan header of the raw data file. This value is different from the actual charge state if you use charge deconvolution, for example, by using Xtract. |
Precursor Charge | Displays the charge state of the precursor ion. |
Precursor Intensity | Displays the intensity of the precursor ion. |
Precursor m/z [Da] | Displays the mass-to-charge ratio of the precursor ion, in daltons. |
Precursor MH+ [Da] | Displays the singly charged mass of the precursor ions, in daltons. |
Quan Info | Displays the status of the validity of this quantification result for the quantification. For reporter ion quantification, it can be one of the following values:
|
Quan Info Details | Displays the reason for rejection when the status in the Quan Info column is Rejected by Method.
|
RT [min] | Displays the retention time when the application observed the peptide, in minutes. |
Scan(s) | Displays the scans from which the actual scan was derived. Usually it is a single scan, but a grouping node might group several scans together. |
SearchID | Displays the order in which the search was submitted to the job queue. |
Spectrum File | Displays the name of the file containing the spectra. |