This chart shows the grouped and ungrouped abundances of a selected protein. In this example, the quantification channels are labeled as study factors rather than as reporter ion masses. Error bars denoting the standard error are displayed above the blue bar that shows the average abundance of the ungrouped abundances. An optional legend at the bottom displays the grouped and ungrouped abundances.
Procedure
- Select the Proteins or Peptide Groups tab in the result file.
- Select the row of the protein or peptide group that interests you.
- Choose View > Quan Channel Values, or select Quan Channel Values
.
The x axis of the chart shows the names of the quantification channels, and the y axis shows the scaled abundance or summed S/N values, depending on whether you chose scaling in the Reporter Ions Quantifier node.
Task | Procedure |
|---|---|
Display the legend | Right-click the Quan Channel Values pane and choose Show Legend. |
Display grouped abundances | Right-click the Quan Channel Values pane and choose Show Grouped Abundances. |
Display ungrouped abundances | Right-click Quan Channel Values pane and choose Show Ungrouped Abundances. |