You can open the FreeStyle application directly in Proteome Discoverer.
Procedure
- Open a result file.
- Select an entry that is directly linked to a RAW file, for example, PMS or MS/MS spectrum information.
- Select Tools > Open FreeStyle to open the application window.
- The FreeStyle application opens.