The following table lists the parameters in the Mass Recalibration view.
Parameter | Description |
|---|---|
ΔM [ppm] | Displays the mass error between the measured mass for a given peptide and the theoretical mass based on the identified sequence, in parts per million. |
RT [min] | Displays the deviation of the experimental mass of the peptide match from the theoretical mass of the peptide. |