You can select a trace or multiple traces to display in the Chromatogram Traces view.
Procedure
- Select the tab of the page in the results file that you want to view.
- Select the row or rows of the traces that you want to view.
- You can select one or multiple traces.
- Choose View > Chromatogram Traces View, or select Chromatogram Traces View
.
- The Chromatogram Traces view opens. It consists of a collapsible pane on the left and a graphical view on the right.
The following figure is an example of a single chromatogram trace.
The following figure is an example of the overlap of multiple selected chromatograms.