The following table lists the parameters in the Bar Charts view.
Command or option | Description |
|---|---|
Options pane | |
Loads the Option pane settings that you saved with the Save command. | |
Saves the settings that you selected in the Options pane.
| |
Resets the option settings in the Options pane to their defaults when you installed the application. | |
Chart Options | |
Chart Type | Determines how the application plots the data.
|
Horizontal Grid Lines | Determines whether the bar chart displays horizontal grid lines and specifies the style of these lines.
|
Axis Options | |
X-Axis Title | Specifies the label for the x axis. Default: The category selected from the Data Source list |
Y-Axis Title | Specifies the label for the y axis. Default: Count |
Y-Axis Type | Specifies the axis type (scale) of the x axis.
|
Axis Title Font | Opens the Font dialog box so that you can specify the font used to denote the labels of the x and y axes. Default: 12-point Arial |
Axis Scale Font | Opens the Font dialog box so that you can specify the font used to denote the scales of the x and y axes. Default: 10-point Arial |
Bar Options | |
Show Rotated Scale | Determines whether the scale labels along the x axis are slightly rotated to the right.
|
X-Axis Scale Angle | Specifies the angle between the label and the x axis for the labels when the Show Rotated Labels parameter is set to True. Range: –90.0 through 90.0 degrees Default: 30 |
Bar Display | Specifies the appearance of the columns or bars.
|
Bar Width | Specifies the relative width of the columns. Range: 0.1–1.0; default: 0.8 |
Show Amount | Determines whether the amount, or count of items, is displayed in the chart.
|
Show Percentage | Determines whether percentage values are displayed above the columns or to the right of the bars.
|
Column Label | Opens the Font dialog box so that you can specify the font of the column labels that appear on top of the bars in the bar chart. Default: 8-point Arial |
Legend Options | |
Show Legend | Determines whether a legend appears and where it appears.
|
Legend Font | Opens the Font dialog box so that you can specify the legend font. Default: 8-point Arial |
Series Options |
|
Show Only Checked Items | Determines whether the chart is compiled from data in all result rows or only data in result rows marked by check marks.
|
Series Color | Specifies the color of the bars. Default: CornflowerBlue |
Data Source | Displays the result category to plot the data from. The categories are as follows: Protein Groups, Proteins, Peptide Groups, PSMs, or MS/MS Spectrum Info. |