In an S plot, the application sorts a single data dimension and plots it against its index. To draw an S plot, select the index data source for the x axis. The index has the same name as the data item name under consideration (PSMs, Proteins, and so forth), followed by the word Index, for example, PSMs Index.
Procedure
- In an open result file, select View > Distribution Charts or select Distribution Charts
.
- Select the Scatter Plots tab in the Report Item Distribution chart.
- From the Data Source list, select the types of data to display.
- In the X Data list, select the data set to display on the x axis.
- For an S plot, select data_set Index.
- In the Y Data list, select the data set to display on the y axis.
- Select Refresh.
See the following figure for an example of an S plot.
Task | Procedure |
|---|---|
Return the settings to the original default settings | In the Options pane, select Factory Defaults. |
Zoom in | Drag the cursor to select the area that you want to enlarge. |
Zoom out | Drag the cursor to the left and select the area that you want to reduce, or right-click the plot and select Zoom Out. |
Return to the default view | Right-click the plot and select Undo All Zoom/Pan. |
Select all the visible data points in the scatter plot | Procedure
|
Clear the visible data points in the scatter plot | Right-click the plot and select Deselect All Visible Points. |
Select a single data point in the scatter plot | Right-click the data point in the scatter plot and select Select Point. |
Deselect a single data point in the scatter plot | Right-click the data point in the scatter plot and select Deselect Point. |
Copy a scatter plot image | Procedure
|
Copy the points in a scatter plot as text | Procedure
|
Save a scatter plot as an image file | Procedure
|
Save the data from the scatter plot in a text file | Procedure
|