Procedure
  1. Select the appropriate neutral loss ion checkboxes in the Neutral Losses area of the Fragments area.
  2. NOTE
    The application only calculates water neutral loss fragments if side chains contain the potential neutral loss groups of aspartic acid, glutamic acid, serine, or threonine. It only calculates ammonia neutral loss fragments if side chains contain the potential neutral loss groups of lysine, asparagine, glutamine, or arginine.
The following table lists the neutral loss ions that you can select.
Neutral loss ions
Ions
Description
–H2O
Neutral loss ion that has lost water (–18 Da)
–NH3
Neutral loss ion that has lost ammonia (–17 Da)
–2H2O
Neutral loss ion that has two water losses (–36 Da)
–2NH3
Neutral loss ion that has lost two ammonia losses (–34 Da)
–CO
Neutral loss ion that has lost (–28 Da)
–HCOOH
Neutral loss ion that has lost (–46 Da)
Modification Loss
Any neutral loss associated with an identified PTM
  1. Select the Neutral Losses tab in the ion table.
The following figure shows the Neutral Losses page of the Peptide Spectrum Match Identification Details view.
Neutral Losses page of the Peptide Spectrum Match Identification Details view
123
No.DescriptionNo.Description
1
Neutral losses
3
Sequence of amino acids that compose the peptide
2
Fragment number