The following table lists the parameters in the Export to MzIdentML dialog box.
Command or Option | Description |
|---|---|
Destination Folder | Specifies the path and name of the MZID file where you save the results of the protein search. |
Checked Proteins Only | Exports all proteins that you selected by putting a check mark next to them on the Proteins page of the result file. Otherwise, the application exports all proteins included in the search results. |
Export mzML File | If checked, the mzML file will be created in addition to the .mzid file. |
Export | Saves the search results in the specified MZID file. |
Close | Closes the Export to MzIdentML dialog box without exporting any search results. |