The following table describes the Export To Excel dialog box parameters.
Command or Option | Description |
|---|---|
Path | Specifies the path and name of the Excel file where you want to save the exported data. |
Items and Associated Tables To Be Exported | Exports the proteins on the selected pages of the result file and determines whether their associated peptides and PSMs are also exported.
|
Options | Determines whether the application exports only selected items or excluded items and whether it automatically opens the Excel file after it finishes exporting the data.
Available only if the Excel application is installed. |
Exports | Exports the selected data. |
Close | Closes the dialog box without exporting any search results. |