The following table describes the Export to Text (Tab Delimited) dialog box parameters.
Command or Option | Description |
|---|---|
Destination Folder | Specifies the path and name of the tab-delimited TXT file where you want to save the exported data. |
Items To Be Exported | Specifies the type of result data that you want to export:
|
Checked Items Only | Exports only the items that you selected. |
Excluded Items Only | Exports only the items that were excluded in the report processing by the Peptide and Protein Filter node. |
Generate R-Friendly Headers | Replaces the special characters in the headers of the columns on the pages of the result file with the appropriate characters instead of dots when you export data to the R Project for Statistical Computing and graphics. |
Export | Saves the selected data in the specified TXT file. |
Close | Closes the Export to Text (Tab Delimited) dialog box without exporting data. |