The following table describes the Export Annotated Spectra dialog box parameters.
Command or Option | Description |
|---|---|
Destination | Specifies the path and name of the PDF file where you want to save the exported annotated spectra. |
Items To Be Exported |
|
Options/Image Size | Specifies the width and height of the spectra.
|
Fragment Match Options | Opens the Options dialog box. For information, see Set global default fragment match options. |
Export | Saves the selected annotated spectra in the specified HTML file. |
Close | Closes the dialog box without exporting any annotated spectra or applying any of the selected options. |