The following figure shows a sample excerpt from a PSMs table in an output HTML file, which shows the annotated spectra for all PSMs.
The following figure shows a sample excerpt of an HTML file, which shows annotated spectra for the PSMs belonging to selected proteins.
The following figure shows a sample excerpt of an HTML file, which shows the annotated spectra of the peptides of selected peptide groups.
The following figure shows the search summary in the HTML file.
The following figure gives an example of the output annotated spectrum in the HTML file.
The annotation includes the following information:
- The path and name of the raw data file
- The numbers of any grouped spectra
- The retention time of the precursor ion, in minutes
- The type of mass spectrometer used to create the data in the raw data file
- The fragmentation method used
- The charge state of the precursor ion
- The mass-to-charge ratio (m/z) of the precursor ion, in daltons
- The protonated monoisotopic mass (MH+), in daltons
- The match tolerance
The following figure gives an example of the output peak list in the HTML file.