The following table describes the Export Spectra dialog box parameters.
Command or option | Description |
|---|---|
Destination Folder | Specifies the path and name of the file where you want to save the exported spectra. |
File Type | Specifies the format of the file to save the exported search results in:
|
Items To Be Exported | Specifies the type of spectra to export. |
From MS/MS Spectrum Info | Exports the spectra extracted from the spectrum file. |
From PSMs | Exports the spectra that identify PSMs. |
From Peptide Groups | Exports the spectra that identify peptide groups and is available when the result file includes peptide groups. |
From Proteins | Exports the spectra that identify proteins and is available when the result file includes proteins. |
Checked Items Only | Exports only the items that you selected on the appropriate page of the result file. |
Export | Saves the selected search results in the specified destination file. |
Close | Closes the dialog box without exporting any spectra or applying any of the selected options. |