Use the following procedure to apply stored filters to a result file or to temporarily override the display of only master proteins on the Proteins page of the result file.
Procedure
- In an open study, create a consensus workflow.
- If it is not already there, drag the Display Settings node to the Post-Processing Nodes pane.
NOTE
Default templates already have the Display Settings node in the Post-Processing Nodes pane. Custom templates do not have it.
- Select the Display Settings node.
- Select the Browse button (...) next to the Filter Set parameter.
- The Filter Set dialog box opens.
NOTE
The default filter set in this dialog box displays only master proteins by default on the Proteins page.
Filter Set (Read Only) dialog box - Select Load File.
- In the Open dialog box, locate the .filterSet file that you want to load, and then select Open.
- In the Parameter Text for Filter Set dialog box, select OK.
- The new filter set replaces the default filter set.
- Run the consensus workflow.