You can check your filter settings by opening the Results Summaries pane.
Procedure
  1. Open a result file.
  2. Select View > Summaries.
  3. In the Result Summaries pane, select the Validation tab.
  4. Select the Filters tab in the upper right corner of the Validation page.
  5. The filters that you set in the Display Filter pane are reflected in the Result Summaries pane.