You can save a result file with a saved filter set.
Procedure
- Select Save to save the filter set in a file on the disk.
- In the Export Filter dialog box, do one of the following:
- Locate the file to save the filter set in.
- Enter the name in the File Name box.
- Select Save.
- The file has a .filterset suffix.
When you save the result file containing the saved filter set and reopen it, the Filter icon, , appears on the tabs of the pages that have filters set to indicated that the results on that page are filtered.