You can disable filters from the Display Filter pane of the result file.
Prerequisites
- You have opened a result file and selected the page you would like to view.
- You have opened the Display Filter pane, see Create a display filter.
Procedure
- In the left pane of the Display Filter pane, select
to turn off the toggle of the filter category that you want to temporarily disable.
- The icon changes to an OFF icon.
- SelectApply.
- (Optional) To enable a filter category, turn on the toggle of the filter, and then select Apply.
- The following figure shows the results when the Peptide Groups filter is temporarily disabled. The disabled filter is denoted by the gold circle.