Display the Found Modifications page - Display the Found Modifications page - Proteome Discoverer 3.1 User Guide - Proteome Discoverer Software - Display the Found Modifications page - Proteome Discoverer Software - Proteome Discoverer Software - topic
Proteome Discoverer 3.1 User Guide
- Product Tree
- Ardia Platform > Software > Proteome Discoverer Software
- Desktop Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- Select the PSMs tab or the Decoy PSMs tab.
- In the Result Table Chooser, select the Unknown Modifications checkbox if it is not already selected.
- Select a row on the PSMs or Decoy PSMs page and select Show Associated Tables.
- Select the Found Modifications tab.