The following table describes the columns of the Decoy Protein Groups page.
Column | Description |
|---|---|
# Peptides | Displays the total number of peptides identified from all included searches for the master protein of the decoy protein group. |
# Proteins | Displays the total number of identified proteins in the decoy protein group. |
# PSMs | Displays the total number of peptide spectrum matches identified from all included searches for the master protein of the decoy protein group. Includes those redundantly identified. |
# Unique Peptides | Displays the number of unique peptide sequences identified from all included searches for the master protein of the decoy protein group. |
Checked | Indicates whether the item is selected. |
Group Description | Displays the name and description of the decoy protein group. |
Protein Group ID | Displays a simple number assigned to the protein group. All protein groups are enumerated. |