The following table describes the columns on the Protein Groups page.
Column | Description |
|---|---|
# Peptides | Displays the total number of peptides identified from all included searches for the master protein of the protein group. |
# Proteins | Displays the total number of identified proteins in the protein group. |
# PSMs | Displays the total number of peptide spectrum matches identified from all included searches for the master protein of the protein group. Includes those redundantly identified. |
# Unique Peptides | Displays the number of unique peptide sequences identified from all included searches for the master protein of the protein group. |
Checked | Indicates whether the item is selected. |
Found in Files | Represents the best confidence of the PSMs of the protein that the application identified in the files:
This column appears only when you include the Data Distributions node in the consensus workflow and set its Show Found in Files parameter to True. |
Found in Fractions | Represents the best confidence of the PSMs of the protein that the application identified in the fractions:
This column appears only when you include the Data Distributions node in the consensus workflow and set its Show Found in Fractions parameter to True. |
Found in Sample Groups | Represents the best confidence of the PSMs of the protein that the application identified in the sample groups:
This column appears only when you include the Data Distributions node in the consensus workflow and set its Show Found in Sample Groups parameter to True. |
Found in Samples | Represents the best confidence of the PSMs of the protein that the application identified in the samples:
This column appears only when you include the Data Distributions node in the consensus workflow and set its Show Found in Samples parameter to True. |
Group Description | Displays a description of the master protein of the protein group. |
Protein Group ID | Displays a simple number assigned to the protein group. All protein groups are enumerated. |