You can apply a default layout or custom layout to a result file during processing.
Procedure
- In an open study, create a consensus workflow. For instructions, see Create a workflow.
- In the Workflow Editor, select the Consensus Workflow tab.
- Drag the Display Settings node to the Post-Processing pane.
- Select the Display Settings node.
- Select the Browse button (...) next to the Layout Definition parameter.
- The Parameter Text for Layout Definition (Readonly) dialog box opens.
- Select Load File.
- In the Open dialog box, locate the layout file that you want to load, and select Open.
- You can find default layouts or layouts that you have already defined in the following folder:
- C:\Users\Public\Documents\Thermo\Proteome Discoverer 3.1\Common Templates\Layouts
- In the Parameter Text for Layout Definition dialog box, select OK.
- The new layout replaces the default layout.
- Run the consensus workflow.