NOTE
You cannot overwrite or save a layout named Original Result Layout.
Do one of the following:
- Select Save to overwrite the existing layout file. By default, this command saves the layout file to the Documents directory.
- Select Save As to save the layout to another layout file. In the Save Layout dialog box, navigate to the location where you want to save the file, and rename the file. The file extension is .pdLayout. Select Save.
- Select Save Common to save the layout file in the common templates directory (C:\Users\Public\Documents\Thermo\Proteome Discoverer 3.1\Common Templates\Layouts).
By default, the Save Layout dialog box opens to the common templates directory. Rename the file. The file extension is .pdLayout.
If a layout should be available for all reports, use this icon to save the layout file to the common template directory.