You can create a layout to apply to your results.
Prerequisites
- You have a result file open.
Procedure
- Select the tab of the page to configure.
- Using the information in the topics under Organizing results, arrange the layout of the page.
- Name the layout by renaming an existing layout (see Manage the contents of the layout list).
- When you change any of the elements of an active layout, the application appends the word modified to the layout name.
- (Optional) Save the layout (see Save a modified layout).
- To restore the original layout after you modify a layout, select Reload Layout.
- The application restores the layout from the corresponding layout file.