You can sort the data in one or more columns of a report by descending order or ascending order.
To display a tooltip that describes the column, point to the column title.
Procedure
- Open the result file.
- Select the header of the column containing the entries to sort.
TIP
To sort the data in multiple columns, hold down the CTRL key while you select the column headers.
- Select the header of the column again, or the arrow icon, to sort the data in the opposite order.
The following figure shows an example of data sorted in descending order in the ΔScore column.