For an individual item on the pages of the result file, you can display the proteins, peptide groups, PSMs, or MS/MS spectrum information associated with it.
Procedure
- On a page of the result file, select the row of the protein group, protein, peptide group, PSM, or spectrum that you are interested in.
- At the bottom left of the page, select Show Associated Tables.
- A table opens at the bottom of the page. In the associated table, the application displays the Proteins page by default.
- The following figure gives an example for an item on the Protein Groups page.
First-level table of items related to protein groups - In the associated table, select the row of the protein group, protein, peptide group, PSM, or spectrum that you are interested in.
- A second-level table opens.
- Select Show Associated Tables again.
Second-level table of items related to protein groups