You can select the columns that are shown in the tables on each page of the result file.
Procedure
- In the upper left of each result page, select Field Chooser
.
- The Field Chooser dialog box opens.
Field Chooser dialog box (PSMs page example) - Do either of the following:
- To display a column, select the checkbox next to the column name.
- To hide a column, clear the selected checkbox.
- The application instantly shows the selected columns and hides the cleared columns.
TIP
You can also remove columns by dragging the column title above the column-title line.
The first few columns of the main table are common to all pages. These columns are described in the following table.
Column | Description |
|---|---|
Select Visible Tables ( | Opens the Select Visible Tables dialog box, so you can select the pages to display in the result file. |
Field Chooser ( | Opens the Field Chooser dialog box, so you can select the columns to display on a page of the result file. |
Index number | Identifies a row. |
Moves the selected row to the top of the results page. | |
Reverses the move of a row to the top of the results page. | |
Checked | Indicates whether the item is selected. |