You can add MSF files or result files to an existing study.
Procedure
  1. In an open study, select Add Files.
  2. The Add Files dialog box opens.
  3. In the Add Files dialog box, select the MSF files or result files that you want to add to the existing results of the study, and then select Open.
  4. To view the MSF files and result files, select the Analysis Results tab.