After you set up the workflow to use for the analysis, you can specify the ratios to report for the quantification and how to group your samples in relation to the specified factor values.
On the Grouping & Quantification page of a study, you can generate custom quantification ratios that do not restrict you to predefined samples and control specifications.
The Grouping & Quantification tab does not appear until you add input files to the analysis.
No. | Description |
|---|---|
1 | Generated Sample Groups area: Displays samples grouped by the values set for the selected study factors. Selecting these variables indicates that you want to group your samples and quantification ratios according to the specified study variables. The grouping of samples affects how identifications are displayed in the distribution maps and the calculation of quantification results. For the latter, the application reports separate area values for each sample or sample group. If the application does not use all available sample group values in at least one ratio, it displays a warning in the Generated Sample Groups area. |
2 | Generated Ratios area: Displays the ratios generated. |
3 | Bulk Ratio Generation area: Displays the study factor values to use as the denominators for multiple generating groups of quantification ratios. |
4 | Manual Ratio Generation area: Contains menus where you can select the numerator and denominator for each quantification ratio. |
5 | Study Variables area: Contains a list of the selectable study factors, or study variables, that you specified in the Study Factors area of the Study Definition page. In this area, you can select the study factors that you want to use to sort samples and quantification ratios into replicate groups and treatment groups.
|
NOTE
The only reason to use a Control is for TMT experiments with multiple plexes. For more information, refer to the Proteome Discoverer Familiarization Guide, exercise five.
NOTE
For more information about the effect of grouping quantification channels based on study variables, see Understanding quantification algorithms.