If you saved a workflow as part of an analysis, you can access the workflow by opening an existing analysis.
Procedure
- Open an existing analysis.
- Select the Workflows tab.
- Do one of the following:
- Select the title bar of the Consensus Step area of the Analysis pane.
- A blue border highlights the Consensus Step area in the Analysis pane.
- Select the title bar of the Processing Step area of the Analysis pane.
- A blue border highlights the Processing Step area in the Analysis pane.
- In the Workflow Editor window, select Open.
- The Open Workflow dialog box opens.
- In the Open Workflow dialog box, locate the .pdProcessingWF file or the .pdConsensusWF file that the custom template is stored in, and then select Open.
- The workflow opens in the Workflow Tree area.
- To correct any template errors, see Correct workflow errors.