Custom workflow templates are templates that you create yourself and store. If you saved a workflow as part of an analysis, you can access the workflow by opening the analysis template.
Procedure
  1. Create an analysis.
  2. Select Open Analysis Template.
  3. The Open Analysis Template dialog box opens.
  4. In the Open Analysis Template dialog box, select the .pdAnalysis file that you want to use as a template.
  5. Select Open.