You can open the workflow that gave the results of a specific results file.
Procedure
- In the Proteome Discoverer application, select Administration > Show Job Queue to open the job queue.
- Select the appropriate search.
- Select Open Study.
- In the study, select the Analysis Results tab.
- Right-click the result of interest and select Show Details.
- The Analysis Sequence Details window displays.
- You can view the Consensus and Processing workflows, Post-Processing Nodes, and Current Workflow Issues for the selected result file.
You can also obtain information about the workflow that generated a result file by using the file’s Search Summary. For information on the Search Summary, see Using the Result Summaries.